Learn More
Thermo Scientific™ TX-150 Swinging Bucket Rotor for Sorvall ST8/ST8R, Megafuge 8/8R and SL8/SL8R Centrifuges
Product Code. 15175233 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
4 x 145 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
15175233 4 x 145 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15175233 Supplier Thermo Scientific™ Supplier No. 75005701
  • ONLINE EXCLUSIVE PRICE
    1385.00€
    1320.00€ / Pack of 1
    Save 65.00€
    4% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Perform a a wide range of clinical and research applications with the Thermo Scientific™ TX-150 Swinging Bucket Rotor, which is highly configurable and convenient.
Perform a wide range of clinical and research applications with the Thermo Scientific™ TX-150 Swinging Bucket Rotor. This rotor accommodates up to 24 x 5/7mL blood collection tubes or 8 x 50mL conical tubes at 4,500rpm/ 3,260 x g, and provides additional flexibility for swing-out processing needs with available adapters. Rotor innovations—from Thermo Scientific™ Auto-Lock™ rotor exchange to Thermo Scientific™ ClickSeal™ biocontainment lids—shorten run set-up time while providing peace-of-mind that samples are secure.
Designed for a broad range of swing-out processing needs at high speed and high capacity with dedicated buckets available for 8 conical tubes capacity or round buckets with ClickSeal biocontainment lids certified by Public Health England, Porton Down, UK.
Secure, push-button Auto-Lock rotor exchange in as little as 3 seconds delivers:
  • Trouble-free rotor installation and removal
  • No tools are required
  • No complicated rotor changing procedures
  • Clear chamber access for cleaning convenience
  • Quick switch between applications, from conical tubes to blood tubes, from microtubes to microplates, and the flexibility to evolve with the changing needs of your laboratory
Ordering Information:
Round buckets (75005702), 50mL Conical buckets (75005703) ClickSeal biocontainment lids (75005707) and replacement O-rings (75005724) are sold separately. A variety of adapters are also available. Contact your sales representative for more information.
TRUSTED_SUSTAINABILITY
Specifications
Description TX-150 Swinging Bucket Rotor
Type Swinging Bucket Rotor
Capacity 4 x 145 mL
Max. RCF 3,260 x g
Max. Speed 4,500 rpm
Radius (Metric) 144 mm
K Factor 12968
Max. Tube Length 100 mm
Max. Tube Diameter 50 mm
For Use With (Equipment) Thermo Scientific Sorvall ST 8, Megafuge 8 and SL 8 Centrifuges
Angle 90°
Biocontainment Yes