Learn More
CORNING GOSSELIN™ Tube Cap, Winged Plugs
Product Code. 15597709
Change view
Click to view available options
Quantity:
3000/Pk.
Unit Size:
Pack of 3000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15597709 3000/Pk. Pack of 3000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15597709 Supplier CORNING GOSSELIN™ Supplier No. BA10038
  • ONLINE EXCLUSIVE PRICE
    78.80€
    52.48€ / Pack of 3000
    Save 26.32€
    33% Off
In Stock
Add to basket
Edge
Add to basket
CORNING GOSSELIN™ Tube Cap, Winged Plugs are produced via injection molding using virgin plastics that comply with current environmental standards and are vertically ribbed for easier grip.
  • All closures and tubes are produced with high-tech equipment using injection molding technology.
  • The raw plastics used are virgin and comply with the current environmental standards.
  • Each production lot is subject to inspection throughout the entire production process according to the standard references, which are reinforced by Gosselin’s own specifications.
  • All closures are vertically ribbed for easier grip.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Ribbed Closure Plug
Material LDPE
Color Natural
No. per Pack 3000/Pk.
No. per Case 15,000/Cs.
Quantity 3000/Pk.
For Use With (Equipment) Tube code: TL75/TC12