Learn More
Vitlab™ Transparent Polypropylene Funnel
Product Code. 11817960
Change view
Click to view available options
Capacity (Metric):
14 mL
32 mL
88 mL
222 mL
342 mL
817 mL
Unit Size:
Each
Pack of 12
Pack of 24
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11817960 32 mL Pack of 24
11827960 14 mL Pack of 24
10622811 222 mL Each
12724098 222 mL Pack of 12
10608972 342 mL Pack of 12
10058540 817 mL Each
10783923 817 mL Pack of 12
10242261 88 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11817960 Supplier Vitlab™ Supplier No. 41294
Only null left
Add to basket
Edge
Add to basket
Use these funnels for a variety of bottling and decanting applications in the food industry. VITLAB™ Transparent Polypropylene Funnels feature a practical handle with loop for hanging. Ideal for powders and liquids.
  • Transparent polypropylene funnel
  • Rapid flow due to a steep 60° angle
Specifications
Diameter (Metric)Top 50mm
Capacity (Metric) 32 mL
Diameter (Metric) Stem 7mm
Material Polypropylene
Height (Metric) 85 mm
Angle 60°
Autoclavable Autoclavable
For Use With (Application) Suitable for food contact
Length (Metric) Stem 43mm