Learn More
Fisherbrand™ Translucent Silicone Tubing
Product Code. 11758705 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Inner:
1 mm
2 mm
3 mm
4 mm
5 mm
6 mm
7 mm
8 mm
10 mm
Diameter (Metric) Outer:
3 mm
6 mm
7 mm
8 mm
9 mm
10 mm
12 mm
14 mm
15 mm
Overall Length (Metric):
5 m
25 m
Unit Size:
25m
5m
Each
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Diameter (Metric) Outer Overall Length (Metric) Unit Size
11758705 1 mm 3 mm 25 m 25m
11773416 10 mm 14 mm 25 m Each
11713855 10 mm 14 mm 5 m Each
11806303 10 mm 15 mm 5 m Each
11743845 1 mm 3 mm 5 m Each
11753845 2 mm 6 mm 5 m Each
11768705 2 mm 6 mm 25 m Each
11778705 3 mm 6 mm 25 m Each
11783845 3 mm 7 mm 5 m Each
11763845 3 mm 6 mm 5 m Each
11876293 4 mm 10 mm 5 m Each
11798705 4 mm 6 mm 25 m Each
11773845 4 mm 6 mm 5 m Each
11763406 5 mm 8 mm 25 m Each
11783855 5 mm 8 mm 5 m 5m
11886293 6 mm 12 mm 5 m Each
11723416 6 mm 9 mm 25 m Each
11793845 6 mm 9 mm 5 m Each
11743416 7 mm 10 mm 25 m Each
11733416 6 mm 12 mm 25 m Each
11703855 7 mm 10 mm 5 m Each
11753416 8 mm 12 mm 25 m Each
11793855 8 mm 12 mm 5 m Each
11763416 8 mm 14 mm 25 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
Product Code. 11758705
In Stock
Add to basket
Edge
Add to basket
Use this tubing with peristaltic pumps. Fisherbrand™ Translucent Silicone Tubing is ideal for pharmaceutical, biotechnology and sterile fluid transfer applications.
  • Translucent silicone tubing
  • FDA compliant (food)
Specifications
Certifications/Compliance FDA, BFR and WRAS approved
Diameter (Metric) Inner 1 mm
Material Silicone
Diameter (Metric) Outer 3 mm
Overall Length (Metric) 25 m
Thickness (Metric) 1 mm
For Use With Peristaltic pumps
For Use With (Application) Pharmaceutical , biotechnology, transfer sterile fluids
Min. Operating Temperature (Metric) -40°C
Sterility Sterile
Color Translucent
Flexibility Flexible
Max. Operating Temperature (Metric) 250°C