Learn More
- Home
- Products
- Tubing
- Rubber and Silicone Tubing
- Fisherbrand™ Translucent Platinum Cured Silicone Tubing
Fisherbrand™ Translucent Platinum Cured Silicone Tubing
Product Code. 11788148 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Inner:
0.5 mm
3.2 mm
4 mm
4.8 mm
6 mm
6.4 mm
8 mm
9.5 mm
9.6 mm
12.8 mm
16 mm
16.0 mm
19.2 mm
Diameter (Metric) Outer:
2.1 mm
4.8 mm
5.6 mm
6.4 mm
8 mm
11.2 mm
11.9 mm
12.8 mm
14.4 mm
15.9 mm
16 mm
17.6 mm
20.8 mm
22.4 mm
25.4 mm
Maximum Pressure:
0.5 bar
1 bar
1.5 bar
1bar
2 bar
30 bar
35 bar
40 bar
Overall Length (Metric):
5 m
25 m
Unit Size:
25m
Each
20 product options available for selection
| Product Code. | Diameter (Metric) Inner | Diameter (Metric) Outer | Maximum Pressure | Overall Length (Metric) | unitSize |
|---|---|---|---|---|---|
| 11788148 | 8 mm | 12.8 mm | 1 bar | 25 m | Each |
| 11728138 | 6.4 mm | 11.2 mm | 1bar | 5 m | Each |
| 11748088 | 0.5 mm | 2.1 mm | 2 bar | 25 m | Each |
| 11788168 | 12.8 mm | 17.6 mm | 0.5 bar | 25 m | Each |
| 11708178 | 16 mm | 20.8 mm | 0.5 bar | 25 m | Each |
| 11798108 | 3.2 mm | 4.8 mm | 0.5 bar | 25 m | Each |
| 11758118 | 4.8 mm | 6.4 mm | 0.5 bar | 25 m | Each |
| 11738118 | 4 mm | 5.6 mm | 0.5 bar | 25 m | Each |
| 11798128 | 6.4 mm | 8 mm | 0.5 bar | 25 m | Each |
| 11763974 | 6 mm | 11.9 mm | 40 bar | 25 m | Each |
| 11768148 | 8 mm | 11.2 mm | 0.5 bar | 25 m | Each |
| 11708158 | 8 mm | 14.4 mm | 1.5 bar | 25 m | Each |
| 11773974 | 8 mm | 14.4 mm | 35 bar | 25 m | Each |
| 11783974 | 9.5 mm | 15.9 mm | 30 bar | 25 m | Each |
| 11728158 | 9.6 mm | 12.8 mm | 0.5 bar | 25 m | Each |
| 11748158 | 9.6 mm | 14.4 mm | 1 bar | 25 m | 25m |
| 11748168 | 9.6 mm | 16 mm | 1 bar | 25 m | Each |
| 11758138 | 6.4 mm | 12.8 mm | 1 bar | 25 m | Each |
| 11728178 | 16.0 mm | 22.4 mm | 1 bar | 25 m | Each |
| 11708188 | 19.2 mm | 25.4 mm | 0.5 bar | 25 m | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
Beschreibung
- Autoclavable
- Non-yellowing
- High gloss finish
- Excellent quality
- Highly hydrophobic
- No peroxide byproducts
- High tear strength as standard
- Extended life in Peristaltic pumps
- Applications include biological, pharmaceutical, biotechnology, aerospace and high temperature work
Spezifikation
Spezifikation
| Certifications/Compliance | FDA, BFR and WRAS approved |
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 8 mm |
| Material | Platinum-cured Silicone |
| Diameter (Metric) Outer | 12.8 mm |
| Overall Length (Metric) | 25 m |
| Thickness (Metric) | 2.4 mm |
| For Use With (Application) | Ideal for biological, pharmaceutical, biotechnology, aerospace, hot work |
| Min. Operating Temperature (Metric) | -40°C |
| Color | Translucent |
| Flexibility | Flexible |
| Maximum Pressure | 1 bar |
| Max. Operating Temperature (Metric) | 200°C |
| Mehr anzeigen Weniger anzeigen |
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?