Learn More
enQuireBio™ TranslationBlocker Human TLR7 / CD287 siRNA
Blocks transcription and translation of TLR7 / CD287 from Human.
Brand: enQuireBio™ QX26-10nmol
Additional Details : Weight : 0.01000kg
Specifications
NP_057646 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
51284 | |
10 nmol | |
Human |
293T Human Embryonic Kidney Cells with Large T Antigen. | |
TLR7 | |
siRNA | |
TLR7-like, TLR7, Toll-like receptor 7, TLR-7 | |
Lyophilized |