missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human PD-L2 / CD273 siRNA
Blocks transcription and translation of PD-L2 / CD273 from Human.
Brand: enQuireBio™ QX8-10nmol
Additional Details : Weight : 0.01000kg
Specifications
NP_079515 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
80380 | |
10 nmol | |
Human |
Hep3b Human Hepatocellular Carinoma Cells, RKO Human Colon Cancer Cells, and Primary Dendritic Cells (DCs) isolated from PBMCs. | |
PDCD1LG2 | |
siRNA | |
PDL2, CD273, Programmed Cell Death 1 Ligand 2, PDCD1L2, B7-DC, B7DC, PD-1 L2, PD-1 Ligand 2, PDCD1 Ligand 2 | |
Lyophilized |