missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human Insulin Receptor siRNA
Blocks transcription and translation of Insulin Receptor from Human.
Brand: enQuireBio™ QX51-2nmol
Additional Details : Weight : 0.01000kg
Specifications
NP_000199 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
3643 | |
2 nmol | |
Human |
HepG2 Human Hepatocellular Carcinoma Cells. | |
Insr | |
siRNA | |
InsR, IR, CD220 | |
Lyophilized |