Learn More
enQuireBio™ TranslationBlocker Human iNOS siRNA
Blocks transcription and translation of iNOS from Human.
Brand: enQuireBio™ QX45-10nmol
Additional Details : Weight : 0.01000kg
Specifications
NP_000616 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
4843 | |
10 nmol | |
Human |
Raji Human Lymphoblastoma Cells. | |
NOS2 | |
siRNA | |
HEP-NOS, NOS2A, NOS, INOS, Nitric oxide synthase, inducible | |
Lyophilized |