missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human FoxP3 siRNA
Blocks transcription and translation of FoxP3 from Human.
Brand: enQuireBio™ QX48-2nmol
Additional Details : Weight : 0.01000kg
Specifications
NP_001107849 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
50943 | |
2 nmol | |
Human |
HCT116-p53wt Human Carcinoma Cells from Colon and MCF7 Human Adenocarcinoma Cells from Breast Cancer. | |
FoxP3 | |
siRNA | |
Forkhead Box P3, Scurfin, IPEX | |
Lyophilized |