Learn More
Alla France™ Precision Thermometers
Product Code. 11756564
Change view
Click to view available options
Temperature Range (Metric):
-10°C to +60°C
-100°C to +30°C
-50°C to +110°C
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Unit Size
11756564 -50°C to +110°C Each
11706574 -10°C to +60°C Each
11832522 -100°C to +30°C Each
11766564 -100°C to +30°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11756564 Supplier Alla France™ Supplier No. 5180T110qp
  • ONLINE EXCLUSIVE PRICE
    22.21€
    12.40€ / Pack of 1
    Save 9.81€
    44% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Use these thermometers for a variety of applications. Alla France™ Precision Thermometers are manufactured using yellow Sodocalcique glass with a diameter of 6 to 7mm.
  • Mercury free : red liquid filling
  • With triangular ring
  • Expansion chamber supports temperatures higher than those of the thermometer
  • The graduation is printed and fused in the glass with a special process
  • Graduation resists to all chemical products except fluoridric acid and hot soda (to a lesser extent) because these 2 products attack the glass
  • Even if you can feel a relief when touching the graduation, the thermometer is not engraved
  • Available in partial or total immersion
  • Individually packed in recyclable sturdy plastic concertina box's, ideal for storing the instrument when not in use
  • Some references are also available in sealed blister by 6 pcs.
  • Systematically delivered with a Precision and Traceability Certificate
Specifications
Bottle Fill Red Liquid
For Use With (Application) General use
Immersion Type Total Immersion
Length (Metric) 305 mm
Material Glass
Thermometer Fill Red Liquid
Product Type Alcohol Thermometer
Max. Temperature (Metric) 110°C
Min. Temperature (Metric) -50°C
Temperature Range (Metric) -50°C to +110°C
Accuracy ±2%