Learn More
BRAND™ Titrating Tube for Titrette™ Bottletop Burette
Includes integrated discharge/recirculation valve.
42.65€ - 77.40€
Specifications
Product Type | Titrating Tube |
---|---|
Includes | Screw cap for tube, discharge/recirculation tube |
Quantity | 1 Ea. |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10798195
|
BRAND™
707526 |
25mL Titrette bottletop burette |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
15663987
|
BRAND™
707525 |
10mL Titrette bottletop burette |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
- For use with Titrette™ Bottle Top Burette
- Screw cap fits onto titrating tube to prevent leakage
- Includes discharge/recirculation valve
Specifications
Titrating Tube | |
Screw cap for tube, discharge/recirculation tube | |
1 Ea. |