Learn More
Cole-Parmer™ Threaded Reducing Nipples
Threaded fittings offer more permanent installation
Brand:  Cole-Parmer™ 06546-27
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11868043
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • Chemically-inert PFA handles most aggressive chemicals
  • Threaded reducing bushing
  • Choose the size and material that meets your application needs
  • Use with our rigid tubing for your high-pressure applications
Specifications
Specifications
Threaded Male Reducer
Autoclavable
1/4 in.
Autoclave or Ethylene Oxide
Translucent
NPT(M) Connection
PFA
1/2 in.
1/2 in. x 1/4 in. NPT(M)