Learn More
Thin Insulating Tubes, Thermo Scientific™
Oxide ceramics tubes resistant to corrosive liquids and gases
Brand:  Thermo Scientific Alfa Aesar 031489.DW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11347518
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Inert to oxidation
  • Not subject to radiation damage
  • Wear resistant and hard
  • With both ends open
Specifications
Specifications
300 mm
1.5mm
2mm