Learn More
Thermo Scientific™ Thermocouple for Portable Muffle Furnaces
For use with Thermolyne Benchtop Muffle Furnaces.
Brand:  Thermo Scientific™ TC48X1A
Additional Details : Weight : 0.01000kg
Product Code. 11807294
  • 96.00€ / Each
Estimated Shipment: 10-05-2024
to see stock.
Add to basket
Description
Description
Thermocouple break protection cuts the power to the heating elements, preventing a thermocouple failure runaway condition.
Specifications
Specifications
Thermolyne Benchtop 1100°C Muffle Furnaces (10-552, 10-552-1)
Thermocouple