Learn More
Thermo Scientific™ Thermocouple for Portable Muffle Furnaces
For use with Thermolyne Benchtop Muffle Furnaces.
Brand: Thermo Scientific™ TC48X1A
Additional Details : Weight : 0.01000kg
Description
Thermocouple break protection cuts the power to the heating elements, preventing a thermocouple failure runaway condition.Specifications
Thermolyne Benchtop 1100°C Muffle Furnaces (10-552, 10-552-1) | |
Thermocouple |