Learn More
Thermo Scientific™ Sorvall™ Adapters for Eight-Place Fixed-Angle Rotor
For use with Thermo Scientific Sorvall Eight-Place Fixed-Angle Rotor 75-003-335
Brand:  Thermo Scientific™ 75003098
Additional Details : Weight : 2.50000kg
Product Code. 10311722
  • 91.05€ / Each
Estimated Shipment: 17-06-2024
Description
Order Info
Sold individually; order correct quantity to equip rotor.
Specifications
Specifications
8-place Fixed angle rotor