Learn More
Thermo Scientific™ High Purity Single Stage Brass Gas Regulators
Recommended for applications where precise flow control is not critical
Brand: Thermo Scientific™ 60180-901
Additional Details : Weight : 4.50000kg
Includes: Diffusion-resistant, diaphragm packless outlet valve to maintain system purity
Description
- For non-corrosive analytical and process applications
- Includes a built-in Smart Purifier Base and Cartridge ensuring 99.99999% gas purity
- Specially designed, convoluted, stainless steel diaphragm provides good regulating performance and maximum purity integrity without the need for a soft seal
- Capable of withstanding an internal vacuum
- All regulators supplied with 1/8 in. stainless steel output adaptor
- Built in check valve allows for cylinder change without contaminating the gas line
Specifications
0 to 200 psig | |
CGA 580 with Triple Filter |