Learn More
Brady™ Thermal Transfer Printable Tags
Description
- Printable
- Polypropylene tags
- For thermal transfer printers
Specifications
Specifications
| Quantity | 1000 |
| Type | Tag |
| Alternate Part Number | THT-33-412-1-EU |
| Color | White |
| Material | Polypropylene |
| Finish | Matte |
| For Use With (Application) | Component and equipment labelling;Data and telecommunications labelling;Electrical labelling;Wire and cable labelling |
| Height (English) | 6 in |
| Height (Metric) | 152.40 mm |
| Shape | Rectangle |
| Show More |