Learn More
Abeba™ The Original 7205 Shoes
Animal fats, oil and fuel resistant. Sanitized™ treated.
Brand:  Abeba™ 7205/42
Additional Details : Weight : 0.76500kg
 View more versions of this product
Product Code. 15682578
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • CE EN ISO 20347:2012 OB FO SRC
  • Smooth leather white with motif
  • Slip resistance outsole sole
  • Sanitized treated
  • Heel strap foldable, adjustable
  • Leather insole
  • Discontinued model
  • Heel cushion
Specifications
Specifications
CE EN ISO 20347:2012 OB FO SRC
Leather
Slip-resistant
Animal fats, oil and fuel
White
42
7205