Learn More
Abeba™ The Original 7091 Shoes
Animal fats, oil and fuel resistant. Sanitized™ treated.
Brand: Abeba™ 7091/43
Additional Details : Weight : 0.80000kg
Description
- CE EN ISO 20347:2012 OB FO SRC
- Smooth leather
- Mint green/White
- Slip resistance outsole sole
- Sanitized treated
- Heel strap foldable, adjustable
- Leather insole
Specifications
CE EN ISO 20347:2012 OB FO SRC | |
Leather | |
Slip-resistant | |
Animal fats, oil and fuel |
Mint Green, White | |
43 | |
7091 |