Learn More
Abeba™ The Original 5081 Shoes
Animal fats, oil and fuel resistant. Sanitized™ treated.
Marke: Abeba™ 5081/39
Weitere Details : Weight : 0.80000kg
Beschreibung
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather
- White/mint
- Slip resistance outsole sole
- Sanitized treated
- Antistatic
- Heel strap foldable, adjustable
- Leather top layer
Spezifikation
CE EN ISO 20347:2012 OB A, FO SRC | |
Leather | |
Slip-resistant | |
Animal fats, oil and fuel |
White | |
39 | |
5081 |