Learn More
Fisherbrand™ Test Tubes, Soda lime, Without Rim, Medium Wall
Product Code. 15802275 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
3 mL
5 mL
7 mL
10 mL
12 mL
18 mL
22 mL
23 mL
29 mL
35 mL
55 mL
Length (Metric):
75 mm
100 mm
125 mm
150 mm
160 mm
180 mm
Diameter (Metric):
10 mm
12 mm
14 mm
16 mm
18 mm
24 mm
Unit Size:
Pack of 100
Pack of 1000
Pack of 50
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Capacity (Metric) Length (Metric) Diameter (Metric) missing translation for 'unitSize'
15802275 29 mL 150 mm 18 mm Pack of 100
15802265 3 mL 75 mm 10 mm Pack of 100
17901132 5 mL 75 mm 12 mm Pack of 1000
17991142 7 mL 100 mm 12 mm Pack of 1000
17911122 10 mL 100 mm 14 mm Pack of 1000
17921122 12 mL 100 mm 16 mm Pack of 1000
17971142 18 mL 125 mm 16 mm Pack of 1000
17951142 22 mL 150 mm 16 mm Pack of 1000
17901122 23 mL 160 mm 16 mm Pack of 1000
17991132 35 mL 180 mm 18 mm Pack of 100
15812275 55 mL 150 mm 24 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 missing translation for 'options'
Código de producto. 15802275
Outlet Corner
Use promo code 28080
Ver todas las promociones
In Stock
Añadir a la cesta
Edge
Añadir a la cesta
Test tubes with round bottom are universal cylindrical laboratory glassware that are used e.g. for chemical reactions, pretreatments or for storing small amounts of liquid.
  • Soda Lime Glass
  • Rimless
  • Disposable
Especificaciones
Certifications/Compliance ISO 4142
Capacity (Metric) 29 mL
Length (Metric) 150 mm
Material Soda Lime Glass
Quantity 100 Pack
Rimmed No
Diameter (Metric) 18 mm
Wall Thickness 1.0 mm
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.