Learn More
Fisherbrand™ Test Tubes, Soda lime, Without Rim, Medium Wall
Shop All Fisherbrand Traceable ProductsSpecifications
Specifications
| Certifications/Compliance | ISO 4142 |
| Capacity (Metric) | 3 mL |
| Length (Metric) | 75 mm |
| Material | Soda Lime Glass |
| Quantity | 100 Pack |
| Rimmed | No |
| Diameter (Metric) | 10 mm |
| Wall Thickness | 1.0 mm |