Learn More
Fisherbrand™ Test Tubes, Borosilicate Glass 3.3, with Beaded Rim, Medium Wall
Código de producto. 17961122 Tienda Fisherbrand Productos
Change view
Click to view available options
Capacity (Metric):
3 mL
5 mL
13 mL
17 mL
21 mL
27 mL
53 mL
57 mL
Length (Metric):
75 mm
100 mm
125 mm
150 mm
160 mm
Diameter (Metric):
10 mm
12 mm
16 mm
18 mm
24 mm
25 mm
Tamaño de la unidad:
Pack of 100
Pack of 50
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Capacity (Metric) Length (Metric) Diameter (Metric) unitSize
17961122 21 mL 160 mm 16 mm Pack of 100
15852285 3 mL 75 mm 10 mm Pack of 100
17921152 5 mL 75 mm 12 mm Pack of 100
17951132 13 mL 100 mm 16 mm Pack of 100
17941132 17 mL 125 mm 16 mm Pack of 100
17901152 21 mL 150 mm 16 mm Pack of 100
17931152 27 mL 150 mm 18 mm Pack of 100
17911152 53 mL 150 mm 24 mm Pack of 50
17971122 57 mL 150 mm 25 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Código de producto. 17961122
Solo quedan null
Añadir a la cesta
Añadir a la cesta
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Test tubes with round bottom are universal cylindrical laboratory glassware that are used e.g. for chemical reactions, pretreatments or for storing small amounts of liquid.
  • Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for this work. Reaction controls, test series and preliminary samples are usually carried out in test tubes due to the small amounts of substance removed.
  • Manufactured from borosilicate glass 3.3
  • Thick-walled and therefore mechanically very resistant
  • With round bottom
TRUSTED_SUSTAINABILITY
Especificaciones
Capacity (Metric) 21 mL
Length (Metric) 160 mm
Material Borosilicate Glass (3.3)
Shape Cylindrical
Sterility Non-sterile
Autoclavable Autoclavable
Graduated No
Diameter (Metric) 16 mm
Product Type Test Tube
Wall Thickness 1.1 mm
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.