Learn More
Velp Scientifica™ Test Tubes for Digestion Units
Product Code. 11715654 Shop All Velp Products
Change view
Click to view available options
Diameter (Metric) Outer:
26 mm
42 mm
48 mm
80 mm
Unit Size:
Each
Pack of 3
Pack of 6
For Use With (Equipment):
DK6/48 and UDK series
Digesters DK 20/26, DKL 42/26 and UDK Distillation Units
UDK Series distillation units
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size For Use With (Equipment)
11715654 42 mm Pack of 3 -
11725654 26 mm Pack of 6 Digesters DK 20/26, DKL 42/26 and UDK Distillation Units
11900114 48 mm Each DK6/48 and UDK series
10777925 80 mm Each UDK Series distillation units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11715654 Supplier Velp Scientifica™ Supplier No. A00000144
  • ONLINE EXCLUSIVE PRICE
    61.15€
    56.50€ / Pack of 3
    Save 4.65€
    7% Off
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
Facilitates the nitrogen analysis and protein determination process. Velp Scientifica™ Test Tubes for Digestion Units are for use with Velp Scientifica™ Kjeldahl Digestion Units.
  • For use in Total Kjeldahl Nitrogen Determination, ammoniacal nitrogen, nitric nitrogen (Devarda), TVBN, sulphites, phenols, volatile acids and cyanides
Specifications
Product Type Test Tubes
Capacity (Metric) 250 mL
Diameter (Metric) Outer 42 mm
Length (Metric) 300 mm
Quantity 3 Tubes