Learn More
Cytiva TempliPhi™ 25 Amplification Sample Kit
For the preparation of circular DNA templates for cycle sequencing, cloning, and transformation
Brand:  Cytiva 29327535
Additional Details : Weight : 0.08000kg
Product Code. 15968872
  • 142.29€ / 25 reactions
Estimated Shipment: 17-06-2024
Description
  • Prepare circular DNA templates for cycle sequencing, cloning, and transformation in 4 to 6 hours
  • Generate microgram quantities of template DNA from picogram amounts of starting material
  • Use amplified DNA directly for cycle sequencing without purification
  • Amplify DNA from bacterial or M13 liquid cultures, colonies, plaques, glycerol stocks, or purified circular (plasmid or M13) DNA
  • Simple protocol reduces time, labor, and consumables needed for template preparation, and workflow enables easy automation
  • For low-to medium-throughput, small DNA vector preparation
Specifications
Specifications
-70°C
Phi29
Amplification Sample Kit
Low to Medium
Preparation of Circular DNA Templates for Cycle Sequencing, Cloning, and Transformation
TempliPhi™
Shipped in Dry Ice