Learn More
Cytiva TempliPhi™ 25 Amplification Sample Kit
For the preparation of circular DNA templates for cycle sequencing, cloning, and transformation
Brand: Cytiva 29327535
Additional Details : Weight : 0.08000kg
Description
- Prepare circular DNA templates for cycle sequencing, cloning, and transformation in 4 to 6 hours
- Generate microgram quantities of template DNA from picogram amounts of starting material
- Use amplified DNA directly for cycle sequencing without purification
- Amplify DNA from bacterial or M13 liquid cultures, colonies, plaques, glycerol stocks, or purified circular (plasmid or M13) DNA
- Simple protocol reduces time, labor, and consumables needed for template preparation, and workflow enables easy automation
- For low-to medium-throughput, small DNA vector preparation
Specifications
-70°C | |
Phi29 | |
Amplification Sample Kit | |
Low to Medium |
Preparation of Circular DNA Templates for Cycle Sequencing, Cloning, and Transformation | |
TempliPhi™ | |
Shipped in Dry Ice |