missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Telmisartan, 99%

An angiotensin II receptor antagonist

72.20€

Chemical Identifiers

CAS 144701-48-4
Formula Weight 514.62g/mol
Synonym BIBR 277; 4′-[(1,4′-Dimethyl-2′-n-propyl[2,6′-bi-1H-benzimidazol]-1′-yl)methyl]biphenyl-2-carboxylic acid
Format Powder
For Use With (Application) An angiotensin II receptor antagonist
Description White
Melting Point 261°C to 263°C
Assay 99%
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15424049
SDS View Documents
Thermo Scientific™
J61441.MD
250 mg
72.20€
250mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

144701-48-4
BIBR 277; 4′-[(1,4′-Dimethyl-2′-n-propyl[2,6′-bi-1H-benzimidazol]-1′-yl)methyl]biphenyl-2-carboxylic acid
An angiotensin II receptor antagonist
261°C to 263°C
514.62g/mol
Powder
White
99%

Specifications

White
An angiotensin II receptor antagonist
C33H30N4O2
BIBR 277; 4′-[(1,4′-Dimethyl-2′-n-propyl[2,6′-bi-1H-benzimidazol]-1′-yl)methyl]biphenyl-2-carboxylic acid
99%
Powder
144701-48-4
514.62g/mol
261°C to 263°C
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.