Learn More
Buerkle™ Telescopic Rod
Product Code. 10182412
Click to view available options
Length (Metric):
115 to 300 cm (Adjustable)
125 to 250 cm (Adjustable)
165 to 450 cm (Adjustable)
175 to 600 cm (Adjustable)
65 to 120 cm (Adjustable)
70 to 180 cm
95 to 280 cm (Adjustable)
Includes:
2 Rods
3 Rods
4 Rods
Unit Size:
Each
Product Code. 10182412
Brand: Buerkle™ 53550450
Edge
Versatile sampling system with exchangeable tools for a wide range of applications.
  • Different tools can be attached to the telescopic rod
  • The tools (angular beaker, pendulum beaker, bottle holder, stainless steel beaker and dip net) are attached to the telescopic rod by a practical snap-in joint
  • Ideal for sampling from baths, tanks, shafts, open water courses etc
  • Sampling depths down to 6 m can be reached with the adjustable telescopic aluminum rod
  • The telescopic rods cannot be rotated radially, which means even better handling
  • The individual rods have been provided with an end stop, this means that one rod cannot accidentally slip out of another rod
  • Telescopic rod 5355-0280, extremely short model – yet extendable up to 280 cm. Fits into every car boot!
  • Springs made of stainless steel
TRUSTED_SUSTAINABILITY
Specifications
Product Type Telescopic Rod
Material Aluminum
Autoclavable Not Autoclavable
Includes 3 Rods
For Use With (Application) Industry and Water Treatment, Ideal for Sampling From Baths, Tanks, Shafts, Open Water Courses, Used with Telescope Sampler
For Use With (Equipment) Bürkle Angled beaker, pendulum beaker, bottle holder, stainless steel beaker and dip net
Length (Metric) 165 to 450 cm (Adjustable)