Learn More
Diversey™ TASKI Jontec Protect High Gloss Floor Emulsion
Protect floor surfaces from scuffs and dirt, especially in high traffic areas. Diversey™ TASKI Jontec Protect High Gloss Floor Emulsion is compatible with most floor types and is ideal for heavy traffic areas where ordinary emulsions require a high level of maintenance.
Brand:  Diversey™ 7513323
Additional Details : Weight : 0.99000kg
Product Code. 10444115
  • 111.00€ / Each
Estimated Shipment: 12-06-2024
Description
  • High-gloss, protective layer
  • Hard, durable, and wear resistant protection
  • Easy to apply, short drying time
  • Slip resistant according to international standards (ASTM D-2047)
  • Easy to repair
  • Enhances the natural appearance of the floor by providing a long lasting high gloss finish
  • Superior resistance to scuff marks and dirt, even under heavy traffic
  • Excellent adhesion on most floor types such as vinyl, synthetic, linoleum, stone, sealed wood, and cork
  • Can be applied with a mop or an applicator
  • Spreads evenly for perfect leveling
  • Responds to most common maintenance methods
  • Ready to use; apply undiluted