Learn More
Fisherbrand™ Tapered PTFE Stir Bars
Product Code. 16255771 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
10 mm
15 mm
20 mm
25 mm
30 mm
35 mm
40 mm
45 mm
50 mm
55 mm
60 mm
70 mm
80 mm
Diameter (Metric):
4 mm
5 mm
7 mm
8 mm
10 mm
Unit Size:
Pack of 10
Pack of 5
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) unitSize
16255771 60 mm 8 mm Pack of 10
16225771 45 mm 8 mm Pack of 10
16265771 70 mm 10 mm Pack of 5
16255761 10 mm 4 mm Pack of 10
16245771 55 mm 8 mm Pack of 10
16235771 50 mm 8 mm Pack of 10
16215771 40 mm 8 mm Pack of 10
16205771 35 mm 8 mm Pack of 10
16275761 20 mm 7 mm Pack of 10
16295761 30 mm 8 mm Pack of 10
16285761 25 mm 8 mm Pack of 10
16265761 15 mm 5 mm Pack of 10
16275771 80 mm 10 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 16255771
Only null left
Add to basket
Add to basket
Provides similar action as oval or pivot ring stirrers. Cowie Technology™ Tapered PTFE Stir Bars are stable and autoclavable for a wide variety of uses.
  • Stable, general-purpose magnetic stir bar
  • Tapered ends provide similar action as oval or pivot ring stirrers
  • Autoclavable
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Coating PTFE
Diameter (Metric) 8 mm
Length (Metric) 60 mm
Quantity 2/Pk.
Shape Tapered Cylindrical
Type Stir Bar