Learn More
Ley Rubber™ Tapered Cork Stoppers
Product Code. 12376457 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric):
19 mm
24 mm
32 mm
Diameter (Metric) Bottom:
11 mm
15 mm
18 mm
Diameter (Metric) Top:
14 mm
18 mm
21 mm
Unit Size:
Pack of 10
Pack of 1000
Pack of 20
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Bottom Diameter (Metric) Top Unit Size
12376457 24 mm 15 mm 18 mm Pack of 20
12396457 32 mm 18 mm 21 mm Pack of 10
12336477 19 mm 11 mm 14 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12376457
  • ONLINE EXCLUSIVE PRICE
    5.59€
    3.79€ / Pack of 20
    Save 1.80€
    32% Off
Estimated Shipment: 22-12-2026
Add to basket
Add to basket
Tapered cork stoppers for use in education, homebrewing and arts and crafts.
  • Range of sizes available
  • Recyclable product
Specifications
For Use With (Application) Education, homebrewing, arts and crafts
Length (Metric) 24 mm
Quantity 20/Pk.
Diameter (Metric) Bottom 15 mm
Diameter (Metric) Top 18 mm
Material Cork
Product Type Cork Stopper
Shape Tapered