Learn More
Brady™ Lockout Labels
Brand:  Brady™ 256198
Additional Details : Weight : 0.04000kg
 View more versions of this product
Product Code. 11746865
  • 26.20€ / 1 pack
Estimated Shipment: 09-07-2024
Specifications
10 Pk.
Red on White
Ne pas fermer
6.3 in.
0.249 mm
-40°F to +212°F
75 mm
7 mm
Tag
Polyester
Lockout Tagout
160 mm
-40°C to +100°C
2.95 in.
160 x 75 mm
Lockout Tagout Tags