Learn More
IKA T 25 Digital Ultra-Turrax™ Immersion Blender
Product Code. 13326309 Shop All IKA Products
Change view
Click to view available options
Speed:
3000 to 25,000 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Unit Size
13326309 3000 to 25,000 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13326309 Supplier IKA Supplier No. 0003725000
  • ONLINE EXCLUSIVE PRICE
    2680.00€
    2539.68€ / Pack of 1
    Save 140.32€
    5% Off
In Stock
Add to basket
Add to basket
High-performance immersion blender. IKA™ T 25 Digital Ultra-Turrax™ offers a wide speed range that enables users to work at high circumferential speeds even with small rotor diameters.
  • Digital speed display
  • Electronic speed control
  • Electronic overload protection
  • Stainless steel dispersing elements can be cleaned quickly and easily
  • Plastic disposable dispersing elements are available in two sizes
  • Error code display
  • Quiet operation
  • EU plug
TRUSTED_SUSTAINABILITY
Specifications
Speed 3000 to 25,000 rpm
Voltage 230 V
Volume Range (Metric) 1 mL to 2 L
Plug Type EU Plug