Learn More
Lenz Laborglasintrumente Stoppers Octagonal Plate, Plastic
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 01401010
Additional Details : Weight : 0.50000kg
Description
- Acc. to DIN 12252
- Form A and E
- Solid or semi-hollow
- Material: Plastic
Spécification
White, Red | |
Polyethylene | |
NS 10/19 |