missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ STET

35.40€ - 51.90€

Specifications

Form Viscous Liquid
Storage Requirements Ambient temperatures
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15423309
SDS View Documents Promotion Details
Thermo Scientific™
J60563.AP
500mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
15433309
SDS View Documents Promotion Details
Thermo Scientific™
J60563.K2
1L This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Viscous Liquid
Ambient temperatures
Videos
SDS
Documents

Documents

Product Certifications