Learn More
Sampling Systems™ SteriWare™ Disposable Mixing Bowl
Ideal for pre-mixing potent drugs and highly coloured substances. Sampling Systems™ SteriWare™ Disposable Mixing Bowl can be used in laboratories and production, as well as small scale work.
Brand: Sampling Systems™ 8017H-30
Additional Details : Weight : 20.39040kg
Description
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- No Cross Contamination
- Can be used straight from the bag
- No Cleaning requierd
- Removes the need for Cleaning Validation
Specifications
Bowl | |
3 L | |
Non-sterile | |
230 mm |
HDPE | |
Yes | |
Natural |