Learn More
Bochem™ Steel Rods
Product Code. 10089020
Change view
Click to view available options
Length (Metric):
1000 mm
1500 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10089020 1000 mm Each
10717171 1500 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10089020 Supplier Bochem™ Supplier No. 5153
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
Made of zincked steel
  • For use with stand bases
  • M10 Thread
  • Zincked steel
Specifications
Material Steel
Length (Metric) 1000 mm
Product Type Rod
For Use With (Application) For Stand Bases
For Use With (Equipment) Stand bases with M10 thread
Quantity 1 Ea.