Learn More
Abeba™ Static Control 1275 Shoes
ESD dissipative and Sanitized™ treated.
Brand: Abeba™ 1275/39
Additional Details : Weight : 1.25000kg
Description
- CE EN ISO 20345:2011 S1 SRA
- Leather with Air-Mesh textile inlays
- Black
- Composite toe cap
- PU midsole
- Slip resistance special rubber outsole sole
- Replaceable acc Wave insole (article no. 3553)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Double velcro fastening
- Reflective material
- Leather with scratch-resistant carbon coating
Specifications
CE EN ISO 20345:2011 S1 SRA | |
Leather | |
Slip-resistant | |
1275 |
Black | |
39 | |
Rubber | |
Composite |