Learn More
Tenak™ Standard Rack for Upright Freezer, 100 mm Box Height, with Boxes
Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.
Brand:  Tenak™ TE23202B
Additional Details : Weight : 3.60000kg
 View more versions of this product
Product Code. 15200704
  • 157.00€ / Each
Estimated Shipment: 23-09-2024
Description
  • Measure the inside height and depth of your freezer when choosing the right rack
  • Racks can be used with all freezer brands
  • Constructed of stainless steel
  • Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications
Specifications
White cardboard boxes, 9 x 9 cell dividers
Boxes (9)
Upright Freezers
3 x 3
Stainless Steel
100 mm (Box)