Learn More
Tenak™ Standard Rack for Microtiterplates, for Upright Freezers, 85 mm Shelf Height
Stainlesss Steel Standard Rack for use in upright freezers is designed to hold microtiterplates.
Brand:  Tenak™ TE27612
Additional Details : Weight : 3.20000kg
 View more versions of this product
Product Code. 15230994
  • 144.00€ / Each
Estimated Shipment: 29-07-2024
Description
  • For use in upright freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of stainless steel
Specifications
Specifications
3 x 7
Stainless Steel
630 x 135 x 259 mm
Microtiterplates (21)
Upright Freezers
85 mm