Learn More
Tenak™ Standard Rack for Microtiterplates, for Upright Freezers, 85 mm Shelf Height
Stainlesss Steel Standard Rack for use in upright freezers is designed to hold microtiterplates.
Brand:  Tenak™ TE27562
Additional Details : Weight : 2.80000kg
 View more versions of this product
Product Code. 15220974
  • 132.00€ / Each
Estimated Shipment: 23-09-2024
Description
  • For use in upright freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of stainless steel
Specifications
Specifications
3 x 6
Stainless Steel
540 x 135 x 259 mm
Microtiterplates (18)
Upright Freezers
85 mm