Learn More
Tenak™ Standard Rack for Microtiterplates, for Upright Freezers, 25 mm Shelf Height
Stainlesss Steel Standard Rack for use in upright freezers is designed to hold microtiterplates.
Brand: Tenak™ TE27580
Additional Details : Weight : 3.40000kg
Description
- For use in upright freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of stainless steel
Specifications
25 mm | |
Upright Freezers | |
6 x 7 |
630 x 135 x 159 mm | |
Microtiterplates (42) | |
Stainless Steel |