Learn More
Tenak™ Standard Rack for Microtiterplates, for Upright Freezers, 25 mm Shelf Height
Stainlesss Steel Standard Rack for use in upright freezers is designed to hold microtiterplates.
Brand:  Tenak™ TE27412
Additional Details : Weight : 3.00000kg
 View more versions of this product
Product Code. 15270914
  • 167.00€ / Each
Estimated Shipment: 20-09-2024
Description
  • For use in upright freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of stainless steel
Specifications
Specifications
25 mm
Upright Freezers
8 x 4
360 x 135 x 212 mm
Microtiterplates (32)