Learn More
Tenak™ Standard Rack for Microtiterplates, for Upright Freezers, 25 mm Shelf Height
Stainlesss Steel Standard Rack for use in upright freezers is designed to hold microtiterplates.
Brand:  Tenak™ TE27584
Additional Details : Weight : 4.20000kg
 View more versions of this product
Product Code. 15200984
  • 233.00€ / Each
Estimated Shipment: 02-10-2024
Description
  • For use in upright freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of stainless steel
Specifications
Specifications
25 mm
Upright Freezers
10 x 7
630 x 135 x 265 mm
Microtiterplates (70)