Learn More
Sartorius™ Standard Flexel™ 3D bags with silicone tubes
Brand: Sartorius™ FXB103364
Additional Details : Weight : 4.00000kg
Specifications
200 L | |
Bags are designed for the preparation, storage and transport of biopharmaceutical solutions, intermediates and final bulk products | |
5/Pk. |
ISO11137 | |
3 Ports | |
Gamma Irradiated |