Learn More
Lauda™ Tube Clip
Material: Stainless Steel; For Use With: 16-27mm tubes
Brand: Lauda™ EZS 032
Additional Details : Weight : 30.00000kg
Specifications
16 to 27 mm tubes | |
Tube Clip | |
Stainless Steel | |
0.5 mm |