Learn More
R&L Enterprises™ Rectangular Retort Stand Bases
Shop All Fisherbrand Products
Click to view available options
Length (Metric) Base:
160 mm
200 mm
250 mm
315 mm
Unit Size:
Each
Specifications
Specifications
| Material | Stainless Steel |
| Length (Metric) Base | 315 mm |
| Width (Metric) Base | 200 mm |
| Includes | With Rubber Feet |
| Product Type | Tripod Plate |
| Color | White |
Product Content Correction
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto