Learn More
Nickel Electro™ Stainless Steel Tongs Bow
Product Code. 11553492 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric):
200 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11553492 200 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11553492
  • ONLINE EXCLUSIVE PRICE
    9.10€
    7.63€ / Pack of 1
    Save 1.47€
    16% Off
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
Laboratory stainless steel bow tong with bright finish.
  • Laboratory bow tong
  • Stainless steel
  • 200 mm
  • With bright finish
Specifications
Product Type Laboratory Bow Tongs
Sterility Non-sterile
Length (Metric) 200 mm
Material Stainless Steel (304)