Learn More
Fisherbrand™ Stainless Steel Sample Dippers
Product Code. 11528122 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
0.12 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11528122 0.12 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11528122
  • ONLINE EXCLUSIVE PRICE
    94.80€
    76.58€ / Pack of 1
    Save 18.22€
    19% Off
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
These stainless steel sampling dippers with tapered sided cups are ideal in laboratories, research, and other environments.
  • Laboratory Sampling Dipper, tapered sided cup, stainless steel 304 spec 18/8 non-magnetic, bright finish
  • Cup style - Tapered
  • Cups have pouring spout
  • Handle argobrazed onto cup for durability and crevice free construction
  • Crevice free cups
  • Autoclaveable
Specifications
Product Type Dip Sampler
Material Stainless Steel
Autoclavable Autoclavable
Capacity (Metric) 0.12 L
Length (Metric) Handle 150 mm