Learn More
RUBIS High Precision Tweezers - SwissTweeze
High Precision Tweezers - Antimagnetic/Anti-acid SS, Perforated Handle
Brand: RUBIS 1G
Additional Details : Weight : 0.02000kg
Description
- High Precision Tweezers
- 100% Made in Switzerland
- Anti-acid/Antimagnetic Stainless Steel
- Perfect alignment
- Very effective
- Reliable
- Excellent chemical resistance
- Swiss Tweeze series
- Perforated handle for enhanced grip
Specifications
Pointed, Ultra Fine | |
4.72 in. | |
120 mm | |
Each |
Straight | |
Anti-acid, Anti-magnetic Stainless Steel | |
High Precision Tweezers | |
Tweezers, Punch |